Learn More
Biomatik Corporation Rat Apoptotic Peptidase Activating Factor 1 (APAF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02537
Encompass_Preferred
Rat Apoptotic Peptidase Activating Factor 1 (APAF1) ELISA Kit for detecting APAF1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EPV5. Gene ID: 78963. Target Synonyms: CED4; Apoptotic Protease Activating Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1861