Learn More
Biomatik Corporation Cattle Aprotinin (AP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02539
Encompass_Preferred
The Cattle Aprotinin (AP) ELISA Kit allows for quantitative determination of Aprotinin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 1.23-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P00974. Cross Activity: No significant cross-reactivity or interference between Aprotinin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1862