Learn More
Biomatik Corporation Human Aquaporin 1, Colton Blood Group (AQP1) ELISA Kit, 96 tests
Catalog No. 501501864
Encompass_Preferred
The Human Aquaporin 1, Colton Blood Group (AQP1) ELISA Kit allows for quantitative determination of Aquaporin 1, Colton Blood Group in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.25-16 ng/mL. Sensitivity: 0.09 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29972. Cross Activity: No significant cross-reactivity or interference between Aquaporin 1, Colton Blood Group and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1864 Supplier Biomatik Corporation Supplier No. EKU02541
The Human Aquaporin 1, Colton Blood Group (AQP1) ELISA Kit allows for quantitative determination of Aquaporin 1, Colton Blood Group in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.25-16 ng/mL. Sensitivity: 0.09 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P29972. Cross Activity: No significant cross-reactivity or interference between Aquaporin 1, Colton Blood Group and analogues was observed. Restriction: For Research Use Only.