Learn More
Biomatik Corporation Human Aquaporin 2, Collecting Duct (AQP2) ELISA Kit, 96 tests
Catalog No. 501501865
Encompass_Preferred
The Human Aquaporin 2, Collecting Duct (AQP2) ELISA Kit allows for quantitative determination of Aquaporin 2, Collecting Duct in Tissue homogenates, Urine and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P41181. Cross Activity: No significant cross-reactivity or interference between Aquaporin 2, Collecting Duct and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1865 Supplier Biomatik Corporation Supplier No. EKU02542
The Human Aquaporin 2, Collecting Duct (AQP2) ELISA Kit allows for quantitative determination of Aquaporin 2, Collecting Duct in Tissue homogenates, Urine and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.117 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P41181. Cross Activity: No significant cross-reactivity or interference between Aquaporin 2, Collecting Duct and analogues was observed. Restriction: For Research Use Only.