Learn More
Biomatik Corporation Rat Aquaporin 2, Collecting Duct (AQP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02543
Encompass_Preferred
Rat Aquaporin 2, Collecting Duct (AQP2) ELISA Kit for detecting AQP2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34080. Gene ID: 25386. Target Synonyms: AQP-CD; WCH-CD; ADH water channel; Aquaporin-CD; Collecting duct water channel protein; Water channel protein for renal collecting duct. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1866