Learn More
Biomatik Corporation Human Aquaporin 3, Gill Blood Group (AQP3) ELISA Kit, 96 tests
Catalog No. 501501867
Encompass_Preferred
The Human Aquaporin 3, Gill Blood Group (AQP3) ELISA Kit allows for quantitative determination of Aquaporin 3, Gill Blood Group in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92482. Cross Activity: No significant cross-reactivity or interference between Aquaporin 3, Gill Blood Group and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1867 Supplier Biomatik Corporation Supplier No. EKU02544
The Human Aquaporin 3, Gill Blood Group (AQP3) ELISA Kit allows for quantitative determination of Aquaporin 3, Gill Blood Group in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92482. Cross Activity: No significant cross-reactivity or interference between Aquaporin 3, Gill Blood Group and analogues was observed. Restriction: For Research Use Only.