Learn More
Biomatik Corporation Mouse Aquaporin 4 (AQP4) ELISA Kit, 96 tests
Catalog No. 501501869
Encompass_Preferred
Mouse Aquaporin 4 (AQP4) ELISA Kit for detecting AQP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55088. Gene ID: 11829. Target Synonyms: HMIWC2; MIWC; WCH4; Mercurial-insensitive water channel. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1869 Supplier Biomatik Corporation Supplier No. EKU02546
Mouse Aquaporin 4 (AQP4) ELISA Kit for detecting AQP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55088. Gene ID: 11829. Target Synonyms: HMIWC2; MIWC; WCH4; Mercurial-insensitive water channel. Restrictions: For Research Use Only. Not for use in diagnostic procedures.