Learn More
Biomatik Corporation Human Aquaporin 9 (AQP9) ELISA Kit, 96 tests
Catalog No. 501501873
Encompass_Preferred
The Human Aquaporin 9 (AQP9) ELISA Kit allows for quantitative determination of Aquaporin 9 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O43315. Cross Activity: No significant cross-reactivity or interference between Aquaporin 9 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1873 Supplier Biomatik Corporation Supplier No. EKU02552
The Human Aquaporin 9 (AQP9) ELISA Kit allows for quantitative determination of Aquaporin 9 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O43315. Cross Activity: No significant cross-reactivity or interference between Aquaporin 9 and analogues was observed. Restriction: For Research Use Only.