Learn More
Biomatik Corporation Human Arginase (ARG) ELISA Kit, 96 tests
Catalog No. 501501876
Encompass_Preferred
The Human Arginase (ARG) ELISA Kit allows for quantitative determination of Arginase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.21 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05089. Cross Activity: No significant cross-reactivity or interference between Arginase and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1876 Supplier Biomatik Corporation Supplier No. EKU02561
The Human Arginase (ARG) ELISA Kit allows for quantitative determination of Arginase in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.21 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05089. Cross Activity: No significant cross-reactivity or interference between Arginase and analogues was observed. Restriction: For Research Use Only.