Learn More
Biomatik Corporation Rat Arginase (ARG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02563
Encompass_Preferred
Rat Arginase (ARG) ELISA Kit for detecting ARG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.1ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07824. Gene ID: 29221. Target Synonyms: ARG1; Arginase I; Liver Arginase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1878