Learn More
Biomatik Corporation Arginine (Arg) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02566
Encompass_Preferred
The Arginine (Arg) ELISA Kit allows for quantitative determination of Arginine in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: All species. Detection Range: 1.23-100 ug/mL. Sensitivity: 0.59 ug/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Arginine and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1881