Learn More
Biomatik Corporation Rat Aromatase (ARO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02572
Encompass_Preferred
Rat Aromatase (ARO) ELISA Kit for detecting ARO in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22443. Target Synonyms: CYP19A1; CYPXIX; ARO; ARO1; CPV1; CYAR; CYP19; P450AROM; P450-AROM; Estrogen synthase; Cytochrome P450,Family 19,Subfamily A,Polypeptide 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1883