Learn More
Biomatik Corporation Human Artemin (ARTN) ELISA Kit, 96 tests
Catalog No. 501501884
Encompass_Preferred
The Human Artemin (ARTN) ELISA Kit allows for quantitative determination of Artemin in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5T4W7. Cross Activity: No significant cross-reactivity or interference between Artemin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1884 Supplier Biomatik Corporation Supplier No. EKU02575
The Human Artemin (ARTN) ELISA Kit allows for quantitative determination of Artemin in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5T4W7. Cross Activity: No significant cross-reactivity or interference between Artemin and analogues was observed. Restriction: For Research Use Only.