Learn More
Biomatik Corporation Human Aryl Hydrocarbon Receptor Nuclear Translocator (ARNT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02578
Encompass_Preferred
The Human Aryl Hydrocarbon Receptor Nuclear Translocator (ARNT) ELISA Kit allows for quantitative determination of Aryl Hydrocarbon Receptor Nuclear Translocator in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P27540. Cross Activity: No significant cross-reactivity or interference between Aryl Hydrocarbon Receptor Nuclear Translocator and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1886