Learn More
Biomatik Corporation Mouse Aspartate Aminotransferase (AST) ELISA Kit, 96 tests
Catalog No. 501501890
Encompass_Preferred
Mouse Aspartate Aminotransferase (AST) ELISA Kit for detecting AST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05201. Gene ID: 14718. Target Synonyms: cCAT; AAT; ASAT; SGOT; GOT1; Cysteine transaminase, cytoplasmic; Transaminase A; Aspartate Transaminase 1,Cytoplasmic; Glutamic-Oxaloacetic Transaminase 1,Soluble. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1890 Supplier Biomatik Corporation Supplier No. EKU02593
Mouse Aspartate Aminotransferase (AST) ELISA Kit for detecting AST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05201. Gene ID: 14718. Target Synonyms: cCAT; AAT; ASAT; SGOT; GOT1; Cysteine transaminase, cytoplasmic; Transaminase A; Aspartate Transaminase 1,Cytoplasmic; Glutamic-Oxaloacetic Transaminase 1,Soluble. Restrictions: For Research Use Only. Not for use in diagnostic procedures.