Learn More
Biomatik Corporation Rat Aspartate Aminotransferase (AST) ELISA Kit, 96 tests
Catalog No. 501501891
Encompass_Preferred
Rat Aspartate Aminotransferase (AST) ELISA Kit for detecting AST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.4ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13221. Gene ID: 24401. Target Synonyms: cCAT; AAT; ASAT; SGOT; GOT1; Cysteine transaminase, cytoplasmic; Transaminase A; Aspartate Transaminase 1,Cytoplasmic; Glutamic-Oxaloacetic Transaminase 1,Soluble. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1891 Supplier Biomatik Corporation Supplier No. EKU02594
Rat Aspartate Aminotransferase (AST) ELISA Kit for detecting AST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.4ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13221. Gene ID: 24401. Target Synonyms: cCAT; AAT; ASAT; SGOT; GOT1; Cysteine transaminase, cytoplasmic; Transaminase A; Aspartate Transaminase 1,Cytoplasmic; Glutamic-Oxaloacetic Transaminase 1,Soluble. Restrictions: For Research Use Only. Not for use in diagnostic procedures.