Learn More
Biomatik Corporation Human Ataxia Telangiectasia Mutated (ATM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02601
Encompass_Preferred
The Human Ataxia Telangiectasia Mutated (ATM) ELISA Kit allows for quantitative determination of Ataxia Telangiectasia Mutated in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q13315. Cross Activity: No significant cross-reactivity or interference between Ataxia Telangiectasia Mutated and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1894