Learn More
Biomatik Corporation Human ATP Binding Cassette Transporter C8 (ABCC8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02619
Encompass_Preferred
The Human ATP Binding Cassette Transporter C8 (ABCC8) ELISA Kit allows for quantitative determination of ATP Binding Cassette Transporter C8 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.25-20 ng/mL. Sensitivity: 0.52 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q09428. Cross Activity: No significant cross-reactivity or interference between ATP Binding Cassette Transporter C8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1897