Learn More
Biomatik Corporation Human ATPase, Ca++ Transporting, Cardiac Muscle, Slow Twitch 2 (ATP2A2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02624
Encompass_Preferred
The Human ATPase, Ca++ Transporting, Cardiac Muscle, Slow Twitch 2 (ATP2A2) ELISA Kit allows for quantitative determination of ATPase, Ca++ Transporting, Cardiac Muscle, Slow Twitch 2 in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.115 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16615. Cross Activity: No significant cross-reactivity or interference between ATPase, Ca++ Transporting, Cardiac Muscle, Slow Twitch 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1899