Learn More
Biomatik Corporation Human ATPase, Cu++ Transporting Beta Polypeptide (ATP7b) ELISA Kit, 96 tests
Catalog No. 501501900
Encompass_Preferred
The Human ATPase, Cu++ Transporting Beta Polypeptide (ATP7b) ELISA Kit allows for quantitative determination of ATPase, Cu++ Transporting Beta Polypeptide in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35670. Cross Activity: No significant cross-reactivity or interference between ATPase, Cu++ Transporting Beta Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1900 Supplier Biomatik Corporation Supplier No. EKU02627
The Human ATPase, Cu++ Transporting Beta Polypeptide (ATP7b) ELISA Kit allows for quantitative determination of ATPase, Cu++ Transporting Beta Polypeptide in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35670. Cross Activity: No significant cross-reactivity or interference between ATPase, Cu++ Transporting Beta Polypeptide and analogues was observed. Restriction: For Research Use Only.