Learn More
Biomatik Corporation Mouse Bactericidal/Permeability Increasing Protein (BPI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02658
Encompass_Preferred
Mouse Bactericidal/Permeability Increasing Protein (BPI) ELISA Kit for detecting BPI in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 14.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q67E05. Gene ID: 329547. Target Synonyms: CAP 57. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1904