Learn More
Biomatik Corporation Human Basic Salivary Proline Rich Protein 1 (PRB1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02662
Encompass_Preferred
The Human Basic Salivary Proline Rich Protein 1 (PRB1) ELISA Kit allows for quantitative determination of Basic Salivary Proline Rich Protein 1 in Saliva and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.59 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04280. Cross Activity: No significant cross-reactivity or interference between Basic Salivary Proline Rich Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1905