Learn More
Biomatik Corporation Mouse B-Cell Activating Factor (BAFF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02665
Encompass_Preferred
Mouse B-Cell Activating Factor (BAFF) ELISA Kit for detecting BAFF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.5-800pg/ml. Sensitivity: 5.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WU72. Gene ID: 24099. Target Synonyms: CD257; TNFSF13B; BLYS; TALL1; THANK; TNFSF20; ZTNF4; B lymphocyte stimulator; Tumor Necrosis Factor Ligand Superfamily Member 13B; Dendritic cell-derived TNF-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1907