Learn More
Biomatik Corporation Rat B-Cell Activating Factor (BAFF) ELISA Kit, 96 tests
Catalog No. 501501908
Encompass_Preferred
Rat B-Cell Activating Factor (BAFF) ELISA Kit for detecting BAFF in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZFD8. Gene ID: 498666. Target Synonyms: CD257; TNFSF13B; BLYS; TALL1; THANK; TNFSF20; ZTNF4; B lymphocyte stimulator; Tumor Necrosis Factor Ligand Superfamily Member 13B; Dendritic cell-derived TNF-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1908 Supplier Biomatik Corporation Supplier No. EKU02666
Rat B-Cell Activating Factor (BAFF) ELISA Kit for detecting BAFF in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78-5000pg/ml. Sensitivity: 27pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZFD8. Gene ID: 498666. Target Synonyms: CD257; TNFSF13B; BLYS; TALL1; THANK; TNFSF20; ZTNF4; B lymphocyte stimulator; Tumor Necrosis Factor Ligand Superfamily Member 13B; Dendritic cell-derived TNF-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.