Learn More
Biomatik Corporation Human B-Cell Leukemia/Lymphoma 2 (Bcl2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02673
Encompass_Preferred
The Human B-Cell Leukemia/Lymphoma 2 (Bcl2) ELISA Kit allows for quantitative determination of B-Cell Leukemia/Lymphoma 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.59 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P10415. Cross Activity: No significant cross-reactivity or interference between B-Cell Leukemia/Lymphoma 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1910