Learn More
Biomatik Corporation Mouse B-Cell Leukemia/Lymphoma 2 (Bcl2) ELISA Kit, 96 tests
Catalog No. 501501911
Encompass_Preferred
Mouse B-Cell Leukemia/Lymphoma 2 (Bcl2) ELISA Kit for detecting Bcl2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10417. Gene ID: 12043. Target Synonyms: B-Cell CLL/Lymphoma 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1911 Supplier Biomatik Corporation Supplier No. EKU02674
Mouse B-Cell Leukemia/Lymphoma 2 (Bcl2) ELISA Kit for detecting Bcl2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10417. Gene ID: 12043. Target Synonyms: B-Cell CLL/Lymphoma 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.