Learn More
Biomatik Corporation Rat Bcl2 Associated X Protein (Bax) ELISA Kit, 96 tests
Catalog No. 501501913
Encompass_Preferred
Rat Bcl2 Associated X Protein (Bax) ELISA Kit for detecting Bax in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O02703. Gene ID: 280730. Target Synonyms: BCL2L4; Bax Zeta; Apoptosis regulator BAX; Bcl-2-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1913 Supplier Biomatik Corporation Supplier No. EKU02681
Rat Bcl2 Associated X Protein (Bax) ELISA Kit for detecting Bax in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O02703. Gene ID: 280730. Target Synonyms: BCL2L4; Bax Zeta; Apoptosis regulator BAX; Bcl-2-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.