Learn More
Biomatik Corporation Rat Beta-2-Microglobulin (b2M) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02693
Encompass_Preferred
Rat Beta-2-Microglobulin (b2M) ELISA Kit for detecting b2M in serum, plasma, urine, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07151. Gene ID: 24223. Target Synonyms: BMG; B2MG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1916