Learn More
Biomatik Corporation Mouse Biglycan (BGN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02722
Encompass_Preferred
Mouse Biglycan (BGN) ELISA Kit for detecting BGN in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.38ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28653. Gene ID: 12111. Target Synonyms: DSPG1; PG-S1; PGI; SLRR1A; Biglycan Proteoglycan; Bone/cartilage proteoglycan I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1923