Learn More
Biomatik Corporation Human B-Lymphocyte Chemoattractant 1 (BLC1) ELISA Kit, 96 tests
Catalog No. 501501926
Encompass_Preferred
The Human B-Lymphocyte Chemoattractant 1 (BLC1) ELISA Kit allows for quantitative determination of B-Lymphocyte Chemoattractant 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q53X90. Cross Activity: No significant cross-reactivity or interference between B-Lymphocyte Chemoattractant 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1926 Supplier Biomatik Corporation Supplier No. EKU02728
The Human B-Lymphocyte Chemoattractant 1 (BLC1) ELISA Kit allows for quantitative determination of B-Lymphocyte Chemoattractant 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q53X90. Cross Activity: No significant cross-reactivity or interference between B-Lymphocyte Chemoattractant 1 and analogues was observed. Restriction: For Research Use Only.