Learn More
Biomatik Corporation Mouse BMP Binding Endothelial Regulator (BMPER) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02734
Encompass_Preferred
Mouse BMP Binding Endothelial Regulator (BMPER) ELISA Kit for detecting BMPER in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8CJ69. Gene ID: 73230. Target Synonyms: Cv2; CRIM3; Crossveinless-2; Bone morphogenetic protein-binding endothelial cell precursor-derived regulator. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1930