Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 1 (BMP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02737
Encompass_Preferred
The Human Bone Morphogenetic Protein 1 (BMP1) ELISA Kit allows for quantitative determination of Bone Morphogenetic Protein 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13497. Cross Activity: No significant cross-reactivity or interference between Bone Morphogenetic Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1933