Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 1 (BMP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02738
Encompass_Preferred
Mouse Bone Morphogenetic Protein 1 (BMP1) ELISA Kit for detecting BMP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P98063. Gene ID: 12153. Target Synonyms: PCOLC; PCP; TLD; mTld; Procollagen C Proteinase; Mammalian tolloid protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1934