Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 15 (BMP15) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02742
Encompass_Preferred
The Human Bone Morphogenetic Protein 15 (BMP15) ELISA Kit allows for quantitative determination of Bone Morphogenetic Protein 15 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O95972. Cross Activity: No significant cross-reactivity or interference between Bone Morphogenetic Protein 15 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1935