Learn More
Biomatik Corporation Cattle Bone Morphogenetic Protein 2 (BMP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02743
Encompass_Preferred
The Cattle Bone Morphogenetic Protein 2 (BMP2) ELISA Kit allows for quantitative determination of Bone Morphogenetic Protein 2 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: A5PJI9. Cross Activity: No significant cross-reactivity or interference between Bone Morphogenetic Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1936