Learn More
Biomatik Corporation Rabbit Bone Morphogenetic Protein 2 (BMP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02749
Encompass_Preferred
Rabbit Bone Morphogenetic Protein 2 (BMP2) ELISA Kit for detecting BMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O46564. Gene ID: 100009349. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1940