Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 6 (BMP6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02761
Encompass_Preferred
Mouse Bone Morphogenetic Protein 6 (BMP6) ELISA Kit for detecting BMP6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20722. Gene ID: 12161. Target Synonyms: VGR; VGR1; Vegetal Related Growth Factor (TGFB-Related) ; VG-1-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1945