Learn More
Biomatik Corporation Mouse Bone Morphogenetic Protein 8B (BMP8B) ELISA Kit, 96 tests
Catalog No. 501501949
Encompass_Preferred
Mouse Bone Morphogenetic Protein 8B (BMP8B) ELISA Kit for detecting BMP8B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55105. Gene ID: 12164. Target Synonyms: BMP8-B; OP2; Osteogenic Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1949 Supplier Biomatik Corporation Supplier No. EKU02767
Mouse Bone Morphogenetic Protein 8B (BMP8B) ELISA Kit for detecting BMP8B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55105. Gene ID: 12164. Target Synonyms: BMP8-B; OP2; Osteogenic Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.