Learn More
Biomatik Corporation Human Bone Sialoprotein (BSP) ELISA Kit, 96 tests
Catalog No. 501501950
Encompass_Preferred
The Human Bone Sialoprotein (BSP) ELISA Kit allows for quantitative determination of Bone Sialoprotein in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8,000 pg/mL. Sensitivity: 51 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P21815. Cross Activity: No significant cross-reactivity or interference between Bone Sialoprotein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1950 Supplier Biomatik Corporation Supplier No. EKU02772
The Human Bone Sialoprotein (BSP) ELISA Kit allows for quantitative determination of Bone Sialoprotein in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8,000 pg/mL. Sensitivity: 51 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P21815. Cross Activity: No significant cross-reactivity or interference between Bone Sialoprotein and analogues was observed. Restriction: For Research Use Only.