Learn More
Biomatik Corporation Human Brain Abundant, Membrane Attached Signal Protein 1 (BASP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02781
Encompass_Preferred
The Human Brain Abundant, Membrane Attached Signal Protein 1 (BASP1) ELISA Kit allows for quantitative determination of Brain Abundant, Membrane Attached Signal Protein 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.114 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P80723. Cross Activity: No significant cross-reactivity or interference between Brain Abundant, Membrane Attached Signal Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1952