Learn More
Biomatik Corporation Guinea Pig Brain Derived Neurotrophic Factor (BDNF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02783
Encompass_Preferred
The Guinea Pig Brain Derived Neurotrophic Factor (BDNF) ELISA Kit allows for quantitative determination of Brain Derived Neurotrophic Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Cavia (Guinea pig). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O70183. Cross Activity: No significant cross-reactivity or interference between Brain Derived Neurotrophic Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1953