Learn More
Biomatik Corporation Human Brain Derived Neurotrophic Factor (BDNF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02784
Encompass_Preferred
The Human Brain Derived Neurotrophic Factor (BDNF) ELISA Kit allows for quantitative determination of Brain Derived Neurotrophic Factor in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 12.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23560. Cross Activity: No significant cross-reactivity or interference between Brain Derived Neurotrophic Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1954