Learn More
Biomatik Corporation Rat Brain Derived Neurotrophic Factor (BDNF) ELISA Kit, 96 tests
Catalog No. 501501956
Encompass_Preferred
Rat Brain Derived Neurotrophic Factor (BDNF) ELISA Kit for detecting BDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23363. Gene ID: 24225. Target Synonyms: Neurotrophin; Abrineurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1956 Supplier Biomatik Corporation Supplier No. EKU02787
Rat Brain Derived Neurotrophic Factor (BDNF) ELISA Kit for detecting BDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23363. Gene ID: 24225. Target Synonyms: Neurotrophin; Abrineurin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.