Learn More
Biomatik Corporation Human Brain Natriuretic Peptide (BNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02794
Encompass_Preferred
The Human Brain Natriuretic Peptide (BNP) ELISA Kit allows for quantitative determination of Brain Natriuretic Peptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 24.69-2000 pg/mL. Sensitivity: 9.67 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P16860. Cross Activity: No significant cross-reactivity or interference between Brain Natriuretic Peptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1958