Learn More
Biomatik Corporation Rat Brain Natriuretic Peptide (BNP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02796
Encompass_Preferred
Rat Brain Natriuretic Peptide (BNP) ELISA Kit for detecting BNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 4.94-400pg/ml. Sensitivity: 1.77pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P13205. Gene ID: 25105. Target Synonyms: GC-B; B-Type Natriuretic Peptide; Ventricular Natriuretic Peptide; Gamma-brain natriuretic peptide; Brain natriuretic peptide 32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1959