Learn More
Biomatik Corporation Human Brain Specific Angiogenesis Inhibitor 1 (BAI1) ELISA Kit, 96 tests
Catalog No. 501501960
Encompass_Preferred
The Human Brain Specific Angiogenesis Inhibitor 1 (BAI1) ELISA Kit allows for quantitative determination of Brain Specific Angiogenesis Inhibitor 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25-2000 pg/mL. Sensitivity: 13.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O14514. Cross Activity: No significant cross-reactivity or interference between Brain Specific Angiogenesis Inhibitor 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1960 Supplier Biomatik Corporation Supplier No. EKU02798
The Human Brain Specific Angiogenesis Inhibitor 1 (BAI1) ELISA Kit allows for quantitative determination of Brain Specific Angiogenesis Inhibitor 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25-2000 pg/mL. Sensitivity: 13.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O14514. Cross Activity: No significant cross-reactivity or interference between Brain Specific Angiogenesis Inhibitor 1 and analogues was observed. Restriction: For Research Use Only.