Learn More
Biomatik Corporation Human Breast Cancer Susceptibility Protein 2 (BRCA2) ELISA Kit, 96 tests
Catalog No. 501501963
Encompass_Preferred
The Human Breast Cancer Susceptibility Protein 2 (BRCA2) ELISA Kit allows for quantitative determination of Breast Cancer Susceptibility Protein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.060 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P51587. Cross Activity: No significant cross-reactivity or interference between Breast Cancer Susceptibility Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1963 Supplier Biomatik Corporation Supplier No. EKU02808
The Human Breast Cancer Susceptibility Protein 2 (BRCA2) ELISA Kit allows for quantitative determination of Breast Cancer Susceptibility Protein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.060 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P51587. Cross Activity: No significant cross-reactivity or interference between Breast Cancer Susceptibility Protein 2 and analogues was observed. Restriction: For Research Use Only.