Learn More
Biomatik Corporation Mouse C Reactive Protein (CRP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02824
Encompass_Preferred
Mouse C Reactive Protein (CRP) ELISA Kit for detecting CRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14847. Gene ID: 12944. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1968