Learn More
Biomatik Corporation Rat C Reactive Protein (CRP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU02826
Encompass_Preferred
Rat C Reactive Protein (CRP) ELISA Kit for detecting CRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48199. Gene ID: 25419. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-1970