Learn More
Biomatik Corporation Human C1q And Tumor Necrosis Factor Related Protein 1 (C1QTNF1) ELISA Kit, 96 tests
Catalog No. 501501972
Encompass_Preferred
The Human C1q And Tumor Necrosis Factor Related Protein 1 (C1QTNF1) ELISA Kit allows for quantitative determination of C1q And Tumor Necrosis Factor Related Protein 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.65 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BXJ1. Cross Activity: No significant cross-reactivity or interference between C1q And Tumor Necrosis Factor Related Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1972 Supplier Biomatik Corporation Supplier No. EKU02828
The Human C1q And Tumor Necrosis Factor Related Protein 1 (C1QTNF1) ELISA Kit allows for quantitative determination of C1q And Tumor Necrosis Factor Related Protein 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.65 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BXJ1. Cross Activity: No significant cross-reactivity or interference between C1q And Tumor Necrosis Factor Related Protein 1 and analogues was observed. Restriction: For Research Use Only.